4-Pentensäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Allylessigsäure
A13991.09
A13991.14
A13991.22
A13991.09EA
61.1
EUR
A13991.09
A13991.14
A13991.22
4-Pentensäure ≥98%
4-Pentensäure
Purity: 98%
CAS No.: 591-80-0
Molecular Formula: H2C=CH(CH2)2CO2H
Formula Weight: 100.12
Flash Point: 89°(192°F)
Refractive Index: 1.4283
Odour: Pungent
Melting Point: -22°
Boiling Point: 187-189°
Solubility: Soluble in alcohol, Slightly soluble in water
Beilstein Reference: 1633696
UN No.: 3265
MDL No.: MFCD00004408
Formel:
C₅H₈O₂ Molecular Weight: 100.12 g/mol Siedepunkt: 187…189 °C Schmelzpunkt: –22 °C Dichte: 0.891 Flammpunkt: 89 °C (192 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004408 CAS-Nummer: 591-80-0 EINECS: 209-732-7 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...