Triallylamin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A12081.0B
A12081.18
A12081.30
A12081.0BEA
295
EUR
A12081.0B
A12081.18
A12081.30
Triallylamin 97%
Triallylamin
Purity: 98%
CAS No.: 102-70-5
Molecular Formula: (H2C=CHCH2)3N
Formula Weight: 137.23
Flash Point: 31°(87°F)
Refractive Index: 1.451
Odour: Amine-like
Melting Point: -70°
Boiling Point: 150-151°
UN No.: 2610
MDL No.: MFCD00026093
Formel:
C₉H₁₅N Siedepunkt: 150…151 °C Schmelzpunkt: –70 °C Dichte: 0,79 Flammpunkt: 31 °C (87 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026093 CAS-Nummer: 102-70-5 EINECS: 203-048-2 UN: 2610 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...