Allylethylether
Lieferant: FLUOROCHEM
Synonyme:
Ethylallylether
001098-25G
001098-5G
FLUO001098-25GEA
0
EUR
FLUO001098-25G
FLUO001098-5G
Allylethylether
Allylethylether
Formel:
C₅H₁₀O Molecular Weight: 86,13 g/mol Siedepunkt: 65 °C (1013 hPa) Dichte: 0,7559 g/cm³ (20 °C) |
MDL:
MFCD00009255 CAS-Nummer: 557-31-3 EINECS: 209-169-7 UN: 2335 ADR: 3,II Merck Index: 13,00290 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...