Nonafluor-2-(trifluormethyl)-2-penten

Lieferant: FLUOROCHEM

Synonyme: 1,1,1,3,4,4,5,5,5-Nonafluor-2-(trifluormethyl)-2-penten, Perfluor(2-methyl-2-penten)

007114-25G 007114-100G
FLUO007114-25GEA 55.4 EUR
FLUO007114-25G FLUO007114-100G
Nonafluor-2-(trifluormethyl)-2-penten
Nonafluor-2-(trifluormethyl)-2-penten
Formel: C₆F₁₂
Molecular Weight: 300.05 g/mol
MDL: MFCD00015724
CAS-Nummer: 1584-03-8
EINECS: 216-436-1
UN: 1993
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR