3-Methyl-3-buten-1-ol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Methyl-1-buten-4-ol, 1-Hydroxy-3-methyl-3-buten
B23398.AC
B23398.AE
B23398.ACEA
24.2
EUR
B23398.AC
B23398.AE
3-Methyl-3-buten-1-ol ≥97%
3-Methyl-3-buten-1-ol
Purity: 97%
CAS No.: 763-32-6
Molecular Formula: CH2=C(CH3)CH2CH2OH
Formula Weight: 86.13
Flash Point: 36°(97°F)
Refractive Index: 1.4330
Boiling Point: 130-133°
Beilstein Reference: 1071239
UN No.: 1987
MDL No.: MFCD00002933
Formel:
C₅H₁₀O Molecular Weight: 86,13 g/mol Siedepunkt: 130…133 °C Schmelzpunkt: –82 °C Dichte: 0,850 Flammpunkt: 36 °C (97 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002933 CAS-Nummer: 763-32-6 EINECS: 212-110-8 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...