2-Methylzimtsäure

Lieferant: MENAI

Synonyme: 3-(o-Methylphenyl)propensäure, 3-(2-Methylphenyl)propensäure, 2-Methylcinnamsäure

DH607.50 DH607.100
MENADH607.50EA 0 EUR
MENADH607.50 MENADH607.100
2-Methylzimtsäure
2-Methylzimtsäure
Formel: C₁₀H₁₀O₂
Molecular Weight: 162.19 g/mol
Siedepunkt: 303
Schmelzpunkt: 175…176 °C
MDL: MFCD00016841
CAS-Nummer: 2373-76-4
EINECS: 219-150-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR