4-Isopropylzimtsäure überwiegend trans ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(p-Isopropylphenyl)propensäure, 4-Isopropylcinnamsäure, 3-(4-Isopropylphenyl)propensäure

A15156.06 A15156.14
A15156.06EA 85.1 EUR
A15156.06 A15156.14
4-Isopropylzimtsäure überwiegend trans ≥98%
4-Isopropylzimtsäure

Purity: 98+% CAS No.: 3368-21-6 Molecular Formula: C12H14O2 Formula Weight: 190.24 Melting Point: 157-161° Beilstein Reference: 2046679

Formel: C₁₂H₁₄O₂
Molecular Weight: 190.24 g/mol
Schmelzpunkt: 157…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016544
CAS-Nummer: 3368-21-6
EINECS: 222-138-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR