4-Isopropylzimtsäure

Lieferant: MENAI
Achtung

Synonyme: 3-(p-Isopropylphenyl)propensäure, 4-Isopropylcinnamsäure, 3-(4-Isopropylphenyl)propensäure

IP50.50 IP50.100
MENAIP50.50EA 0 EUR
MENAIP50.50 MENAIP50.100
4-Isopropylzimtsäure
4-Isopropylzimtsäure
Formel: C₁₂H₁₄O₂
Molecular Weight: 190.24 g/mol
MDL: MFCD00016544
CAS-Nummer: 3368-21-6
EINECS: 222-138-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR