4-(Trifluormethyl)zimtsäure

Lieferant: FLUOROCHEM
Achtung

007615-5G 007615-25G
FLUO007615-5GEA 0 EUR
FLUO007615-5G FLUO007615-25G
4-(Trifluormethyl)zimtsäure
4-(Trifluormethyl)zimtsäure
Formel: C₁₀H₇F₃O₂
MDL: MFCD00002696
CAS-Nummer: 2062-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR