trans-3-Bromzimtsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(3-Bromphenyl)propensäure, 3-(m-Bromphenyl)propensäure, 3-Bromcinnamsäure
A16035.06
A16035.14
A16035.22
A16035.06EA
56
EUR
A16035.06
A16035.14
A16035.22
trans-3-Bromzimtsäure ≥98%
trans-3-Bromzimtsäure
Purity: 98+%
CAS No.: 14473-91-7
Molecular Formula: C9H7BrO2
Formula Weight: 227.06
Odour: Odourless
Melting Point: 178-182°
Beilstein Reference: 2084230
MDL No.: MFCD00004382
Formel:
C₉H₇BrO₂ Molecular Weight: 227,06 g/mol Siedepunkt: 323 °C (1013 hPa) Schmelzpunkt: 178…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004382 CAS-Nummer: 14473-91-7 EINECS: 251-267-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...