4-Bromzimtsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-(4-Bromphenyl)propensäure, 3-(p-Bromphenyl)propensäure, 4-Bromcinnamsäure
001109-25G
001109-5G
001109-100G
FLUO001109-25GEA
42.9
EUR
FLUO001109-25G
FLUO001109-5G
FLUO001109-100G
4-Bromzimtsäure
4-Bromzimtsäure
Formel:
C₉H₇BrO₂ Molecular Weight: 227,06 g/mol |
MDL:
MFCD00004394 CAS-Nummer: 1200-07-3 EINECS: 214-850-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...