5-Brom-2-methoxyzimtsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
5-Brom-2-methoxycinnamsäure, 3-(5-Brom-2-methoxyphenyl)propensäure
011098-25G
011098-100G
011098-5G
FLUO011098-25GEA
0
EUR
FLUO011098-25G
FLUO011098-100G
FLUO011098-5G
5-Brom-2-methoxyzimtsäure
5-Brom-2-methoxyzimtsäure
Formel:
C₁₀H₉BrO₃ |
MDL:
MFCD00017596 CAS-Nummer: 40803-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...