5-Brom-2-methoxyzimtsäure

Lieferant: MENAI
Achtung

Synonyme: 5-Brom-2-methoxycinnamsäure, 3-(5-Brom-2-methoxyphenyl)propensäure

IP14.50 IP14.100
MENAIP14.50EA 0 EUR
MENAIP14.50 MENAIP14.100
5-Brom-2-methoxyzimtsäure
5-Brom-2-methoxyzimtsäure
Formel: C₁₀H₉BrO₃
Molecular Weight: 257,08 g/mol
MDL: MFCD00017596
CAS-Nummer: 40803-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR