4-Brom-2-fluorzimtsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(4-Brom-2-fluorphenyl)propensäure, 4-Brom-2-fluorcinnamsäure
004225-5G
004225-25G
FLUO004225-5GEA
31.1
EUR
FLUO004225-5G
FLUO004225-25G
4-Brom-2-fluorzimtsäure
4-Brom-2-fluorzimtsäure
Formel:
C₉H₆BrFO₂ Molecular Weight: 245.05 g/mol Siedepunkt: 322 Schmelzpunkt: 219…223 °C |
MDL:
MFCD00143267 CAS-Nummer: 149947-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...