5-Brom-2-fluorzimtsäure

Lieferant: FLUOROCHEM

004226-5G 004226-1G
FLUO004226-5GEA 97.2 EUR
FLUO004226-5G FLUO004226-1G
5-Brom-2-fluorzimtsäure
5-Brom-2-fluorzimtsäure
Formel: C₉H₆BrFO₂
MDL: MFCD00143427
CAS-Nummer: 202865-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR