2-Chlorzimtsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-Chlorcinnamsäure, 3-(o-Chlorphenyl)propensäure, 3-(2-Chlorphenyl)propensäure
001142-100G
001142-500G
FLUO001142-100GEA
133
EUR
FLUO001142-100G
FLUO001142-500G
2-Chlorzimtsäure
2-Chlorzimtsäure
Formel:
C₉H₇ClO₂ Molecular Weight: 182.61 g/mol Siedepunkt: 311 Schmelzpunkt: 210…211 °C |
MDL:
MFCD00004372 CAS-Nummer: 3752-25-8 EINECS: 223-154-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...