4-Chlor-3-nitrozimtsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(4-Chloro-3-nitrophenyl)prop-2-enoic acid, 3-(4-Chloro-3-nitrophenyl)acrylic acid

A14954.06 A14954.22
A14954.06EA 23.4 EUR
A14954.06 A14954.22
4-Chlor-3-nitrozimtsäure ≥98%
4-Chlor-3-nitrozimtsäure

Purity: 98% CAS No.: 20797-48-2 Molecular Formula: C9H6ClNO4 Formula Weight: 227.61 Melting Point: 186-188° MDL No.: MFCD00063311

Formel: C₉H₆ClNO₄
Molecular Weight: 227,601 g/mol
Schmelzpunkt: 186…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063311
CAS-Nummer: 20797-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR