2-Chlor-3,6-difluorzimtsäure

Lieferant: FLUOROCHEM

005602-5G 005602-1G
FLUO005602-5GEA 488 EUR
FLUO005602-5G FLUO005602-1G
2-Chlor-3,6-difluorzimtsäure
2-Chlor-3,6-difluorzimtsäure
Formel: C₉H₅ClF₂O₂
Molecular Weight: 218,59 g/mol
MDL: MFCD01631464
CAS-Nummer: 261762-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR