3-Chlor-2-fluorzimtsäure
Lieferant: FLUOROCHEM
Synonyme:
3-Chlor-2-fluorcinnamsäure, 3-(3-Chlor-2-fluorphenyl)propensäure
005636-1G
005636-5G
FLUO005636-1GEA
79.1
EUR
FLUO005636-1G
FLUO005636-5G
3-Chlor-2-fluorzimtsäure
3-Chlor-2-fluorzimtsäure
Formel:
C₉H₆ClFO₂ Molecular Weight: 200,6 g/mol |
MDL:
MFCD01631392 CAS-Nummer: 261762-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...