3-Chlor-4-fluorzimtsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(3-Chlor-4-fluorphenyl)propensäure, 3-Chlor-4-fluorcinnamsäure

005637-1G 005637-5G
FLUO005637-1GEA 39.9 EUR
FLUO005637-1G FLUO005637-5G
3-Chlor-4-fluorzimtsäure
3-Chlor-4-fluorzimtsäure
Formel: C₉H₆ClFO₂
MDL: MFCD01631393
CAS-Nummer: 155814-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR