4-Chlor-2-fluorzimtsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-(4-Chlor-2-fluorphenyl)propensäure, 4-Chlor-2-fluorcinnamsäure
013800-5G
013800-25G
FLUO013800-5GEA
94.1
EUR
FLUO013800-5G
FLUO013800-25G
4-Chlor-2-fluorzimtsäure
4-Chlor-2-fluorzimtsäure
Formel:
C₉H₆ClFO₂ Molecular Weight: 200.6 g/mol |
MDL:
MFCD00143287 CAS-Nummer: 202982-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...