4-Chlor-3-fluorzimtsäure

Lieferant: CHEMPUR

FL13272.POR 013272-5G
CHMPFL13272.POREA 0 EUR
CHMPFL13272.POR FLUO013272-5G
4-Chlor-3-fluorzimtsäure
4-Chlor-3-fluorzimtsäure
Formel: C₉H₆ClFO₂
Molecular Weight: 200.6 g/mol
MDL: MFCD00143294
CAS-Nummer: 202982-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR