trans-2-Nitrozimtsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Nitrocinnamsäure, 3-(2-Nitrophenyl)propensäure, 3-(o-Nitrophenyl)propensäure

A13171.14 A13171.22 A13171.36
A13171.14EA 58.7 EUR
A13171.14 A13171.22 A13171.36
trans-2-Nitrozimtsäure ≥98%
trans-2-Nitrozimtsäure

Purity: 98% CAS No.: 1013-96-3 Molecular Formula: C9H7NO4 Formula Weight: 193.16 Odour: Odourless Melting Point: 245-249° Beilstein Reference: 2211209 MDL No.: MFCD00007189

Formel: C₉H₇NO₄
Molecular Weight: 193,16 g/mol
Schmelzpunkt: 245…249 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007189
CAS-Nummer: 1013-96-3
EINECS: 210-309-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR