3-Nitrozimtsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(3-Nitrophenyl)acrylic acid

002996-25G 002996-100G
FLUO002996-25GEA 75.6 EUR
FLUO002996-25G FLUO002996-100G
3-Nitrozimtsäure
3-Nitrozimtsäure
Formel: C₉H₇NO₄
Molecular Weight: 193,160 g/mol
MDL: MFCD00007277
CAS-Nummer: 555-68-0
EINECS: 209-104-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR