4'-Nitrozimtsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: trans-4-Nitrocinnamic acid, trans-3-(4-Nitrophenyl)propenoic acid

A12990.14 A12990.22
A12990.14EA 48.6 EUR
A12990.14 A12990.22
4'-Nitrozimtsäure ≥98%
4'-Nitrozimtsäure

Purity: 98+% CAS No.: 882-06-4 Molecular Formula: C9H7NO4 Formula Weight: 193.16 Odour: Odourless Melting Point: ca 290° dec. Beilstein Reference: 1369659 MDL No.: MFCD00007381

Formel: C₉H₇NO₄
Molecular Weight: 193,16 g/mol
Schmelzpunkt: ∼ 290 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007381
CAS-Nummer: 882-06-4
EINECS: 212-924-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR