trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
trans-Ferulasäure, 4-Hydroxy-3-methoxyzimtsäure
A13890.14
A13890.22
A13890.36
A13890.14EA
55.6
EUR
A13890.14
A13890.22
A13890.36
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥99%
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
Purity: 99%
CAS No.: 537-98-4
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 170-175°
Beilstein Reference: 1371483
Merck Reference: 14,4062
MDL No.: MFCD00004400
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,18 g/mol Schmelzpunkt: 170…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004400 CAS-Nummer: 537-98-4 EINECS: 208-679-7 Merck Index: 14,4062 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...