2,3,4-Trifluorzimtsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2,3,4-Trifluorcinnamsäure, 3-(2,3,4-Trifluorphenyl)propensäure

007970-1G 007970-5G
FLUO007970-1GEA 87.9 EUR
FLUO007970-1G FLUO007970-5G
2,3,4-Trifluorzimtsäure
2,3,4-Trifluorzimtsäure
Formel: C₉H₅F₃O₂
Molecular Weight: 202.13 g/mol
MDL: MFCD00083552
CAS-Nummer: 207742-85-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR