2,3,5-Trifluorzimtsäure
Lieferant: FLUOROCHEM
007971-1G
007971-5G
FLUO007971-1GEA
55.4
EUR
FLUO007971-1G
FLUO007971-5G
2,3,5-Trifluorzimtsäure
2,3,5-Trifluorzimtsäure
Formel:
C₉H₅F₃O₂ Molecular Weight: 202.13 g/mol |
CAS-Nummer:
237761-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...