2,3,6-Trifluorzimtsäure

Lieferant: FLUOROCHEM

Synonyme: 2,3,6-Trifluorcinnamsäure, 3-(2,3,6-Trifluorphenyl)propensäure

007972-1G 007972-5G
FLUO007972-1GEA 209 EUR
FLUO007972-1G FLUO007972-5G
2,3,6-Trifluorzimtsäure
2,3,6-Trifluorzimtsäure
Formel: C₉H₅F₃O₂
Molecular Weight: 202.13 g/mol
MDL: MFCD00061213
CAS-Nummer: 230295-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR