2,4,5-Trifluorzimtsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2,4,5-Trifluorcinnamsäure, 3-(2,4,5-Trifluorphenyl)propensäure
007973-1G
007973-5G
FLUO007973-1GEA
39.9
EUR
FLUO007973-1G
FLUO007973-5G
2,4,5-Trifluorzimtsäure
2,4,5-Trifluorzimtsäure
Formel:
C₉H₅F₃O₂ Molecular Weight: 202.13 g/mol |
MDL:
MFCD00061211 CAS-Nummer: 247170-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...