trans-2,5-Difluorzimtsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-(2,5-Difluorphenyl)propensäure, 2,5-Difluorcinnamsäure

004806-1G 004806-5G 004806-10G 004806-25G
FLUO004806-1GEA 31.1 EUR
FLUO004806-1G FLUO004806-5G FLUO004806-10G FLUO004806-25G
trans-2,5-Difluorzimtsäure
trans-2,5-Difluorzimtsäure
Formel: C₉H₆F₂O₂
Molecular Weight: 184.14 g/mol
MDL: MFCD00010318
CAS-Nummer: 112898-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR