trans-2,6-Difluorzimtsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-(2,6-Difluorphenyl)propensäure, 2,6-Difluorcinnamsäure

004807-1G 004807-5G 004807.10G 004807-25G
FLUO004807-1GEA 31.1 EUR
FLUO004807-1G FLUO004807-5G FLUO004807.10G FLUO004807-25G
trans-2,6-Difluorzimtsäure
trans-2,6-Difluorzimtsäure
Formel: C₉H₆F₂O₂
Molecular Weight: 184.14 g/mol
MDL: MFCD00010319
CAS-Nummer: 102082-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR