2-Fluorzimtsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(2-Fluorophenyl)propenoic acid

001440-25G 001440-100G
FLUO001440-25GEA 42.9 EUR
FLUO001440-25G FLUO001440-100G
2-Fluorzimtsäure
2-Fluorzimtsäure
Formel: C₉H₇FO₂
Molecular Weight: 166,15 g/mol
MDL: MFCD00004370
CAS-Nummer: 451-69-4
EINECS: 207-195-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR