2-Fluorzimtsäure

Lieferant: MENAI

IP43.25 IP43.50
MENAIP43.25EA 0 EUR
MENAIP43.25 MENAIP43.50
2-Fluorzimtsäure
2-Fluorzimtsäure
Formel: C₉H₇FO₂
MDL: MFCD00004370
CAS-Nummer: 451-69-4
EINECS: 207-195-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR