3,4,5-Trifluorzimtsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3,4,5-Trifluorcinnamsäure, 3-(3,4,5-Trifluorphenyl)propensäure

007975-1G 007975-5G
FLUO007975-1GEA 39.9 EUR
FLUO007975-1G FLUO007975-5G
3,4,5-Trifluorzimtsäure
3,4,5-Trifluorzimtsäure
Formel: C₉H₅F₃O₂
Molecular Weight: 202.13 g/mol
MDL: MFCD00083529
CAS-Nummer: 152152-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR