trans-3,4-Difluorzimtsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-(3,4-Difluorphenyl)propensäure, 3,4-Difluorcinnamsäure

004808-1G 004808-5G 004808-25G
FLUO004808-1GEA 30.5 EUR
FLUO004808-1G FLUO004808-5G FLUO004808-25G
trans-3,4-Difluorzimtsäure
trans-3,4-Difluorzimtsäure
Formel: C₉H₆F₂O₂
Molecular Weight: 184.14 g/mol
MDL: MFCD00010320
CAS-Nummer: 112897-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR