trans-3,5-Difluorzimtsäure

Lieferant: FLUOROCHEM
Achtung

004809-1G 004809-25G 004809-5G
FLUO004809-1GEA 31.1 EUR
FLUO004809-1G FLUO004809-25G FLUO004809-5G
trans-3,5-Difluorzimtsäure
trans-3,5-Difluorzimtsäure
Formel: C₉H₆F₂O₂
Schmelzpunkt: 199…201 °C
MDL: MFCD00010321
CAS-Nummer: 147700-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR