4-Fluorzimtsäure 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(p-Fluorphenyl)propensäure, 4-Fluorcinnamsäure, 3-(4-Fluorphenyl)propensäure
207110050
207110250
ACRO207110050EA
36.3
EUR
ACRO207110050
ACRO207110250
4-Fluorzimtsäure 99%
4-Fluorzimtsäure
Formel:
C₉H₇FO₂ Molecular Weight: 166.15 g/mol Schmelzpunkt: 207…210 °C |
MDL:
MFCD00004395 CAS-Nummer: 459-32-5 EINECS: 207-288-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 207 to 210 °C |
HPLC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...