4-Fluorzimtsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(p-Fluorphenyl)propensäure, 4-Fluorcinnamsäure, 3-(4-Fluorphenyl)propensäure

A16707.06 A16707.14 A16707.22
A16707.06EA 38.5 EUR
A16707.06 A16707.14 A16707.22
4-Fluorzimtsäure ≥98%
4-Fluorzimtsäure

Purity: 98+% CAS No.: 459-32-5 Molecular Formula: C9H7FO2 Formula Weight: 166.15 Odour: Odourless Melting Point: 208-210° Beilstein Reference: 2613441 MDL No.: MFCD00004395

Formel: C₉H₇FO₂
Molecular Weight: 166,15 g/mol
Schmelzpunkt: 208…210 °C
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004395
CAS-Nummer: 459-32-5
EINECS: 207-288-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR