Zimtalkohol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Phenylallylalkohol, Cinnamylalkohol, trans-3-Phenylprop-2-en-1-ol, Styron, trans-3-Phenyl-2-propen-1-ol, trans-3-Hydroxy-1-phenylpropen, trans-3-Phenylallylalkohol

A13025.0B A13025.30
A13025.0BEA 88.7 EUR
A13025.0B A13025.30
Zimtalkohol ≥98%
Zimtalkohol

Purity: 98% CAS No.: 104-54-1 Molecular Formula: C9H10O Formula Weight: 134.18 Flash Point: 126°(258°F) Odour: Pleasant Boiling Point: 257-258° Beilstein Reference: 1903999 Merck Reference: 14,2302 MDL No.: MFCD00002921

Formel: C₉H₁₀O
Molecular Weight: 134.18 g/mol
Siedepunkt: 257…258 °C
Schmelzpunkt: 32…35 °C
Dichte: 1.044
Flammpunkt: 126 °C (258 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002921
CAS-Nummer: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR