5-Chlor-1-pentin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L02835.06
L02835.14
L02835.06EA
60.9
EUR
L02835.06
L02835.14
5-Chlor-1-pentin ≥98%
5-Chlor-1-pentin
Purity: 98%
CAS No.: 14267-92-6
Molecular Formula: C5H7Cl
Formula Weight: 102.56
Storage Temperature: 2°C to 8°C
Flash Point: 15°(59°F)
Refractive Index: 1.4440
Odour: Recognizable
Boiling Point: 113°
Beilstein Reference: 1736710
UN No.: 1993
MDL No.: MFCD00001014
Formel:
C₅H₇Cl Molecular Weight: 102,56 g/mol Siedepunkt: 113 °C Dichte: 0,968 Flammpunkt: 15 °C (59 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001014 CAS-Nummer: 14267-92-6 EINECS: 238-159-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...