3-Hexin-2-on ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Hex-3-yn-2-one

L05315.04 L05315.09
L05315.04EA 80.9 EUR
L05315.04 L05315.09
3-Hexin-2-on ≥97%
3-Hexin-2-on

Purity: 97% CAS No.: 1679-36-3 Molecular Formula: C6H8O Formula Weight: 96.13 Flash Point: 39°(102°F) Refractive Index: 1.4410 Boiling Point: 67-68°/50mm Beilstein Reference: 1699613 UN No.: 1992 MDL No.: MFCD00041627

Formel: C₆H₈O
Molecular Weight: 96,13 g/mol
Siedepunkt: 67…68 °C (50 mmHg)
Dichte: 0,880
Flammpunkt: 39 °C (102 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041627
CAS-Nummer: 1679-36-3
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR