2-Butinsäureethylester, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Ethyl-2-butinoat

M-Clarity™ Qualitätsstufe = MQ200

425117-10G 425117-1G
SIAL425117-10GEA 146 EUR
SIAL425117-10G SIAL425117-1G
2-Butinsäureethylester, Sigma-Aldrich®
2-Butinsäureethylester
Formel: C₆H₈O₂
Molecular Weight: 112,13 g/mol
MDL: MFCD00015182
CAS-Nummer: 4341-76-8
EINECS: 224-395-6
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR