2-Butinsäure ≥98%

Lieferant: Thermo Scientific
Gefahr

A15542.03 A15542.06 A15542.14
A15542.03EA 51.8 EUR
A15542.03 A15542.06 A15542.14
2-Butinsäure ≥98%
2-Butinsäure

Purity: 98% CAS No.: 590-93-2 Molecular Formula: C4H4O2 Formula Weight: 84.07 Storage Temperature: 2°C to 8°C Melting Point: 75-79° Boiling Point: 200-203° Beilstein Reference: 1740205 UN No.: 3261 MDL No.: MFCD00004363

Formel: C₄H₄O₂
Molecular Weight: 84,07 g/mol
Siedepunkt: 200…203 °C
Schmelzpunkt: 75…79 °C
Dichte: 0,964
MDL: MFCD00004363
CAS-Nummer: 590-93-2
EINECS: 209-695-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR