2-Pentin-1-ol ≥98%

Lieferant: Thermo Scientific
Achtung

A12999.03 A12999.06 A12999.14
A12999.03EA 22.5 EUR
A12999.03 A12999.06 A12999.14
2-Pentin-1-ol ≥98%
2-Pentin-1-ol

Purity: 98% CAS No.: 6261-22-9 Molecular Formula: C5H8O Formula Weight: 84.12 Storage Temperature: 2°C to 8°C Flash Point: 58°(136°F) Refractive Index: 1.4520 Odour: Recognizable Boiling Point: 154° Beilstein Reference: 1734007 UN No.: 1987 MDL No.: MFCD00040915

Formel: C₅H₈O
Molecular Weight: 84,12 g/mol
Siedepunkt: 154 °C
Schmelzpunkt: –0,2 °C
Dichte: 0,910
Flammpunkt: 58 °C (136 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00040915
CAS-Nummer: 6261-22-9
EINECS: 228-411-2
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR