4-Pentin-1-ol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Pentyn-5-ol
A10405.06
A10405.14
A10405.22
A10405.06EA
59.5
EUR
A10405.06
A10405.14
A10405.22
4-Pentin-1-ol ≥98%
4-Pentin-1-ol
Purity: 98%
CAS No.: 5390-04-5
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 62°(143°F)
Refractive Index: 1.4450
Boiling Point: 153-155°
Beilstein Reference: 1736712
MDL No.: MFCD00002974
Formel:
C₅H₈O Molecular Weight: 84.12 g/mol Siedepunkt: 153…155 °C Dichte: 0.907 Flammpunkt: 62 °C (143 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002974 CAS-Nummer: 5390-04-5 EINECS: 226-383-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...