3-Chlor-3-methyl-1-butin 96%
Lieferant: Thermo Scientific
|
Gefahr
|
378010010
378010050
378010250
ACRO378010010EA
29.3
EUR
ACRO378010010
ACRO378010050
ACRO378010250
3-Chlor-3-methyl-1-butin 96%
3-Chlor-3-methyl-1-butin
Formel:
C₅H₇Cl Molecular Weight: 102,56 g/mol Flammpunkt: 4 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00190221 CAS-Nummer: 1111-97-3 EINECS: 214-188-9 UN: 1993 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to straw yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4190 (20°C, 589 nm) |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...