3-Chlor-3-methyl-1-butin 95%
Lieferant: Thermo Scientific
|
Gefahr
|
B25252.03
B25252.06
B25252.14
B25252.03EA
26.8
EUR
B25252.03
B25252.06
B25252.14
3-Chlor-3-methyl-1-butin 95%
3-Chlor-3-methyl-1-butin
Purity: 95%
CAS No.: 1111-97-3
Molecular Formula: C5H7Cl
Formula Weight: 102.57
Storage Temperature: 2°C to 8°C
Flash Point: 4°(40°F)
Refractive Index: 1.4178
Odour: Recognizable
Melting Point: -61°
Boiling Point: 73-75°
UN No.: 1993
MDL No.: MFCD00190221
Formel:
C₅H₇Cl Molecular Weight: 102,56 g/mol Siedepunkt: 73…75 °C Schmelzpunkt: –61 °C Dichte: 0,913 Flammpunkt: 4 °C (40 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00190221 CAS-Nummer: 1111-97-3 EINECS: 214-188-9 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...