3-Chlor-3-methyl-1-butin

Lieferant: FLUOROCHEM

009094-10G 009094-50G
FLUO009094-10GEA 0 EUR
FLUO009094-10G FLUO009094-50G
3-Chlor-3-methyl-1-butin
3-Chlor-3-methyl-1-butin
Formel: C₅H₇Cl
Molecular Weight: 102,56 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00190221
CAS-Nummer: 1111-97-3
EINECS: 214-188-9
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR