3-Chlor-3-methyl-1-butin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
301345-25G
301345-5G
SIAL301345-25GEA
354
EUR
SIAL301345-25G
SIAL301345-5G
3-Chlor-3-methyl-1-butin, Sigma-Aldrich®
3-Chlor-3-methyl-1-butin
Formel:
C₅H₇Cl Molecular Weight: 102,56 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00190221 CAS-Nummer: 1111-97-3 EINECS: 214-188-9 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...